Assay Details
| Gene Symbol: LOC4343793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791482.2 | XP_015646968.1 | 16 - 17 | 5192 | 75 | |
| GenBank mRNA | AK120222.1 | - | 13 - 14 | 2172 | 75 | |
| HQ123580.1 | - | 16 - 17 | 4792 | 75 | ||
More Information
Set Membership: |
|

LOC4343793
Genomic Map