Assay Details
| Gene Symbol: LOC4343793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791482.2 | XP_015646968.1 | 8 - 9 | 4418 | 143 | |
| GenBank mRNA | AK120222.1 | - | 5 - 6 | 1398 | 143 | |
| HQ123580.1 | - | 8 - 9 | 4018 | 143 | ||
More Information
Set Membership: |
|

LOC4343793
Genomic Map