Assay Details
| Gene Symbol: LOC4343793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791482.2 | XP_015646968.1 | 10 - 11 | 4627 | 63 | |
| GenBank mRNA | AK120222.1 | - | 7 - 8 | 1607 | 63 | |
| HQ123580.1 | - | 10 - 11 | 4227 | 63 | ||
More Information
Set Membership: |
|

LOC4343793
Genomic Map