Assay Details
| Gene Symbol: LOC4343793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791482.2 | XP_015646968.1 | 12 - 13 | 4802 | 96 | |
| GenBank mRNA | AK120222.1 | - | 9 - 10 | 1782 | 96 | |
| HQ123580.1 | - | 12 - 13 | 4402 | 96 | ||
More Information
Set Membership: |
|

LOC4343793
Genomic Map