Assay Details
| Gene Symbol: LOC4344071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791886.1 | XP_015647372.1 | 2 - 3 | 359 | 63 | |
| GenBank mRNA | AK059154.1 | - | 1 - 2 | 105 | 63 | |
| AK119728.1 | - | 2 - 3 | 324 | 63 | ||
| AK122129.1 | - | 2 - 3 | 325 | 63 | ||
| BT129187.1 | - | 2 - 3 | 327 | 63 | ||
| BT130576.1 | - | 2 - 3 | 318 | 63 | ||
| BT131414.1 | - | 2 - 3 | 316 | 63 | ||
More Information
Set Membership: |
|

LOC4344071
Genomic Map