Assay Details
| Gene Symbol: LOC4344572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794951.2 | XP_015650437.1 | 1 - 2 | 165 | 72 | |
| GenBank mRNA | AK059350.1 | - | 1 - 2 | 157 | 72 | |
| AK099242.1 | - | 1 - 2 | 104 | 72 | ||
| AK121348.1 | - | 1 - 2 | 157 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794951.2 transcripts.
Set Membership: |
|

LOC4344572
Genomic Map