Assay Details
| Gene Symbol: LOC4344572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794951.2 | XP_015650437.1 | 6 - 7 | 1517 | 58 | |
| GenBank mRNA | AK059350.1 | - | 6 - 7 | 1509 | 58 | |
| AK099242.1 | - | 6 - 7 | 1456 | 58 | ||
| AK121348.1 | - | 6 - 7 | 1509 | 58 | ||
More Information
Set Membership: |
|

LOC4344572
Genomic Map