Assay Details
| Gene Symbol: LOC4344572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794951.2 | XP_015650437.1 | 7 - 8 | 1615 | 137 | |
| GenBank mRNA | AK059350.1 | - | 7 - 8 | 1607 | 137 | |
| AK099242.1 | - | 7 - 8 | 1554 | 137 | ||
| AK121348.1 | - | 7 - 8 | 1607 | 137 | ||
More Information
Set Membership: |
|

LOC4344572
Genomic Map