Assay Details
| Gene Symbol: LOC4344940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794914.2 | XP_015650400.1 | 1 - 2 | 154 | 84 | |
| GenBank mRNA | AK106835.1 | - | 1 - 2 | 49 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794914.2 transcripts.
Set Membership: |
|

LOC4344940
Genomic Map