Assay Details
| Gene Symbol: LOC4345330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795375.1 | XP_015650861.1 | 1 - 2 | 230 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795375.1 transcripts.
Set Membership: |
|

LOC4345330
Genomic Map