Assay Details
| Gene Symbol: LOC4345332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792602.2 | XP_015648088.1 | 5 - 6 | 1417 | 127 | |
| GenBank mRNA | AK111861.1 | - | 5 - 6 | 1422 | 127 | |
More Information
Set Membership: |
|

LOC4345332
Genomic Map