Assay Details
| Gene Symbol: LOC4345636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795144.2 | XP_015650630.1 | 10 - 11 | 1166 | 61 | |
| GenBank mRNA | AK101681.1 | - | 10 - 11 | 1075 | 61 | |
| AK102785.1 | - | 10 - 11 | 1122 | 61 | ||
More Information
Set Membership: |
|

LOC4345636
Genomic Map