Assay Details
| Gene Symbol: LOC4345636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795144.2 | XP_015650630.1 | 8 - 9 | 998 | 85 | |
| GenBank mRNA | AK101681.1 | - | 8 - 9 | 907 | 85 | |
| AK102785.1 | - | 8 - 9 | 954 | 85 | ||
More Information
Set Membership: |
|

LOC4345636
Genomic Map