Assay Details
| Gene Symbol: LOC4346676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755429.2 | XP_015610915.1 | 1 - 2 | 120 | 134 | |
| GenBank mRNA | AK066404.1 | - | 1 - 2 | 119 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755429.2 transcripts.
Set Membership: |
|

LOC4346676
Genomic Map