Assay Details
| Gene Symbol: LOC4346676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755429.2 | XP_015610915.1 | 5 - 6 | 456 | 125 | |
| GenBank mRNA | AK066404.1 | - | 5 - 6 | 455 | 125 | |
More Information
Set Membership: |
|

LOC4346676
Genomic Map