Assay Details
| Gene Symbol: LOC4326932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765834.2 | XP_015621320.1 | 3 - 4 | 646 | 97 | |
| XM_015765836.2 | XP_015621322.1 | 3 - 4 | 641 | 97 | ||
| XM_015765837.2 | XP_015621323.1 | 3 - 4 | 646 | 97 | ||
| XM_015765838.2 | XP_015621324.1 | 3 - 4 | 646 | 97 | ||
| GenBank mRNA | AK065414.1 | - | 3 - 4 | 599 | 97 | |
More Information
Set Membership: |
|

LOC4326932
Genomic Map