Assay Details
| Gene Symbol: LOC4326552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758567.2 | XP_015614053.1 | 11 - 12 | 1163 | 90 | |
| GenBank mRNA | AK073858.1 | - | 11 - 12 | 1168 | 90 | |
More Information
Set Membership: |
|

LOC4326552
Genomic Map