Assay Details
| Gene Symbol: LOC4327961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771937.1 | XP_015627423.1 | 5 - 6 | 820 | 66 | |
| XM_015771945.2 | XP_015627431.1 | 5 - 6 | 728 | 66 | ||
| XM_015771954.2 | XP_015627440.1 | 5 - 6 | 760 | 66 | ||
| XM_015771962.2 | XP_015627448.1 | 5 - 6 | 709 | 66 | ||
| GenBank mRNA | AK102132.1 | - | 5 - 6 | 735 | 66 | |
More Information
Set Membership: |
|

LOC4327961
Genomic Map