Assay Details
| Gene Symbol: LOC4327961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771937.1 | XP_015627423.1 | 7 - 8 | 1056 | 69 | |
| XM_015771945.2 | XP_015627431.1 | 7 - 8 | 964 | 69 | ||
| XM_015771954.2 | XP_015627440.1 | 7 - 8 | 996 | 69 | ||
| XM_015771962.2 | XP_015627448.1 | 7 - 8 | 945 | 69 | ||
| GenBank mRNA | AK102132.1 | - | 7 - 8 | 971 | 69 | |
More Information
Set Membership: |
|

LOC4327961
Genomic Map