Assay Details
| Gene Symbol: LOC4328912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769994.2 | XP_015625480.1 | 2 - 3 | 520 | 109 | |
| GenBank mRNA | AK065296.1 | - | 2 - 3 | 384 | 109 | |
| AK100312.1 | - | 2 - 3 | 511 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769994.2 transcripts.
Set Membership: |
|

LOC4328912
Genomic Map