Assay Details
| Gene Symbol: LOC4328912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769994.2 | XP_015625480.1 | 4 - 5 | 1262 | 84 | |
| GenBank mRNA | AK065296.1 | - | 4 - 5 | 1126 | 84 | |
| AK100312.1 | - | 4 - 5 | 1253 | 84 | ||
More Information
Set Membership: |
|

LOC4328912
Genomic Map