Assay Details
| Gene Symbol: LOC4328912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769994.2 | XP_015625480.1 | 6 - 7 | 1652 | 73 | |
| GenBank mRNA | AK065296.1 | - | 6 - 7 | 1516 | 73 | |
| AK100312.1 | - | 5 - 6 | 1728 | 73 | ||
| AK112041.1 | - | 1 - 2 | 69 | 73 | ||
More Information
Set Membership: |
|

LOC4328912
Genomic Map