Assay Details
| Gene Symbol: LOC4328912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769994.2 | XP_015625480.1 | 7 - 8 | 1892 | 119 | |
| GenBank mRNA | AK100312.1 | - | 6 - 7 | 1968 | 119 | |
| AK112041.1 | - | 2 - 3 | 309 | 119 | ||
More Information
Set Membership: |
|

LOC4328912
Genomic Map