Assay Details
| Gene Symbol: LOC4345965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794764.2 | XP_015650250.1 | 6 - 7 | 1528 | 99 | |
| XM_015794765.1 | XP_015650251.1 | 6 - 7 | 1528 | 99 | ||
| GenBank mRNA | AK065218.1 | - | 6 - 7 | 1537 | 99 | |
| AK065539.1 | - | 6 - 7 | 1519 | 99 | ||
More Information
Set Membership: |
|

LOC4345965
Genomic Map