Assay Details
Gene Symbol: LOC4351982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763228.2 | XP_015618714.1 | 3 - 4 | 612 | 116 | |
GenBank mRNA | AK070295.1 | - | 3 - 4 | 619 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
