Assay Details
| Gene Symbol: LOC4352265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765087.2 | XP_015620573.1 | 2 - 3 | 482 | 54 | |
| GenBank mRNA | AK073582.1 | - | 2 - 3 | 453 | 54 | |
More Information
Set Membership: |
|

LOC4352265
Genomic Map