Assay Details
| Gene Symbol: LOC4352265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765087.2 | XP_015620573.1 | 5 - 6 | 952 | 125 | |
| GenBank mRNA | AK073582.1 | - | 5 - 6 | 923 | 125 | |
More Information
Set Membership: |
|

LOC4352265
Genomic Map