Assay Details
| Gene Symbol: LOC9270404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761711.2 | XP_015617197.1 | 19 - 20 | 2433 | 114 | |
| XM_015761712.2 | XP_015617198.1 | 19 - 20 | 2433 | 114 | ||
| XM_015761713.2 | XP_015617199.1 | 18 - 19 | 2286 | 114 | ||
| GenBank mRNA | AK067123.1 | - | 16 - 17 | 2220 | 114 | |
More Information
Set Membership: |
|

LOC9270404
Genomic Map