Assay Details
| Gene Symbol: LOC9270404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761711.2 | XP_015617197.1 | 23 - 24 | 2854 | 106 | |
| XM_015761712.2 | XP_015617198.1 | 23 - 24 | 2854 | 106 | ||
| XM_015761713.2 | XP_015617199.1 | 22 - 23 | 2707 | 106 | ||
| GenBank mRNA | AK067123.1 | - | 20 - 21 | 2641 | 106 | |
More Information
Set Membership: |
|

LOC9270404
Genomic Map