Assay Details
| Gene Symbol: LOC9270404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761711.2 | XP_015617197.1 | 28 - 29 | 3247 | 113 | |
| XM_015761713.2 | XP_015617199.1 | 27 - 28 | 3100 | 113 | ||
| GenBank mRNA | AK059481.1 | - | 3 - 4 | 197 | 113 | |
| AK067123.1 | - | 25 - 26 | 3034 | 113 | ||
More Information
Set Membership: |
|

LOC9270404
Genomic Map