Assay Details
| Gene Symbol: LOC4329132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772104.2 | XP_015627590.1 | 2 - 3 | 434 | 122 | |
| GenBank mRNA | AK058414.1 | - | 2 - 3 | 274 | 122 | |
More Information
Set Membership: |
|

LOC4329132
Genomic Map