Assay Details
| Gene Symbol: LOC4329132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772104.2 | XP_015627590.1 | 4 - 5 | 612 | 74 | |
| GenBank mRNA | AK058414.1 | - | 4 - 5 | 452 | 74 | |
More Information
Set Membership: |
|

LOC4329132
Genomic Map