Assay Details
| Gene Symbol: LOC4329132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772104.2 | XP_015627590.1 | 6 - 7 | 745 | 62 | |
| GenBank mRNA | AK058414.1 | - | 6 - 7 | 585 | 62 | |
| BT130065.1 | - | 1 - 2 | 52 | 62 | ||
More Information
Set Membership: |
|

LOC4329132
Genomic Map