Assay Details
| Gene Symbol: LOC4332612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775486.2 | XP_015630972.1 | 1 - 2 | 262 | 57 | |
| GenBank mRNA | AK243493.1 | - | 1 - 2 | 107 | 57 | |
More Information
Set Membership: |
|

LOC4332612
Genomic Map