Assay Details
| Gene Symbol: LOC4332665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775610.2 | XP_015631096.1 | 10 - 11 | 1077 | 82 | |
| XM_015775611.2 | XP_015631097.1 | 11 - 12 | 1204 | 82 | ||
| XM_015775612.2 | XP_015631098.1 | 10 - 11 | 1595 | 82 | ||
| XM_015775613.2 | XP_015631099.1 | 10 - 11 | 1085 | 82 | ||
| GenBank mRNA | AK111920.1 | - | 7 - 8 | 610 | 82 | |
| AK111929.1 | - | 9 - 10 | 909 | 82 | ||
| AY256847.1 | - | 10 - 11 | 1030 | 82 | ||
| EU703794.1 | - | 9 - 10 | 909 | 82 | ||
More Information
Set Membership: |
|

LOC4332665
Genomic Map