Assay Details
| Gene Symbol: LOC4332665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775610.2 | XP_015631096.1 | 13 - 14 | 1354 | 67 | |
| XM_015775611.2 | XP_015631097.1 | 14 - 15 | 1481 | 67 | ||
| XM_015775612.2 | XP_015631098.1 | 13 - 14 | 1872 | 67 | ||
| XM_015775613.2 | XP_015631099.1 | 13 - 14 | 1362 | 67 | ||
| GenBank mRNA | AK111752.1 | - | 13 - 14 | 1878 | 67 | |
| AK111920.1 | - | 10 - 11 | 887 | 67 | ||
| AK111929.1 | - | 12 - 13 | 1186 | 67 | ||
| AY256847.1 | - | 13 - 14 | 1307 | 67 | ||
| EU703794.1 | - | 12 - 13 | 1186 | 67 | ||
More Information
Set Membership: |
|

LOC4332665
Genomic Map