Assay Details
| Gene Symbol: LOC4332665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775610.2 | XP_015631096.1 | 3 - 4 | 421 | 132 | |
| GenBank mRNA | AK111929.1 | - | 2 - 3 | 253 | 132 | |
| AY256847.1 | - | 3 - 4 | 374 | 132 | ||
| EU703794.1 | - | 2 - 3 | 253 | 132 | ||
| EU807984.1 | - | 2 - 3 | 253 | 132 | ||
More Information
Set Membership: |
|

LOC4332665
Genomic Map