Assay Details
| Gene Symbol: LOC4332665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775610.2 | XP_015631096.1 | 5 - 6 | 606 | 123 | |
| XM_015775611.2 | XP_015631097.1 | 6 - 7 | 733 | 123 | ||
| XM_015775612.2 | XP_015631098.1 | 5 - 6 | 1124 | 123 | ||
| XM_015775613.2 | XP_015631099.1 | 5 - 6 | 614 | 123 | ||
| GenBank mRNA | AK111752.1 | - | 5 - 6 | 1130 | 123 | |
| AK111920.1 | - | 2 - 3 | 139 | 123 | ||
| AK111929.1 | - | 4 - 5 | 438 | 123 | ||
| AY256847.1 | - | 5 - 6 | 559 | 123 | ||
| EU703794.1 | - | 4 - 5 | 438 | 123 | ||
| EU807984.1 | - | 4 - 5 | 438 | 123 | ||
More Information
Set Membership: |
|

LOC4332665
Genomic Map