Assay Details
| Gene Symbol: LOC4332665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775610.2 | XP_015631096.1 | 6 - 7 | 687 | 121 | |
| XM_015775611.2 | XP_015631097.1 | 7 - 8 | 814 | 121 | ||
| XM_015775612.2 | XP_015631098.1 | 6 - 7 | 1205 | 121 | ||
| XM_015775613.2 | XP_015631099.1 | 6 - 7 | 695 | 121 | ||
| GenBank mRNA | AK111752.1 | - | 6 - 7 | 1211 | 121 | |
| AK111920.1 | - | 3 - 4 | 220 | 121 | ||
| AK111929.1 | - | 5 - 6 | 519 | 121 | ||
| AY256847.1 | - | 6 - 7 | 640 | 121 | ||
| EU703794.1 | - | 5 - 6 | 519 | 121 | ||
| EU807984.1 | - | 5 - 6 | 519 | 121 | ||
More Information
Set Membership: |
|

LOC4332665
Genomic Map