Assay Details
| Gene Symbol: LOC4334537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775598.2 | XP_015631084.1 | 13 - 14 | 2175 | 135 | |
| GenBank mRNA | AK061963.1 | - | 8 - 9 | 1019 | 135 | |
| AK072463.1 | - | 13 - 14 | 2081 | 135 | ||
| EF065605.1 | - | 13 - 14 | 2175 | 135 | ||
More Information
Set Membership: |
|

LOC4334537
Genomic Map