Assay Details
| Gene Symbol: LOC4334537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775598.2 | XP_015631084.1 | 1 - 2 | 379 | 74 | |
| GenBank mRNA | AK072463.1 | - | 1 - 2 | 285 | 74 | |
| EF065605.1 | - | 1 - 2 | 379 | 74 | ||
More Information
Set Membership: |
|

LOC4334537
Genomic Map