Assay Details
| Gene Symbol: LOC4336232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781577.2 | XP_015637063.1 | 12 - 13 | 2195 | 78 | |
| GenBank mRNA | AK120849.1 | - | 12 - 13 | 1996 | 78 | |
More Information
Set Membership: |
|

LOC4336232
Genomic Map