Assay Details
| Gene Symbol: LOC4340890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788773.2 | XP_015644259.2 | 10 - 11 | 1180 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4340890| Gene Symbol: LOC4340890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788773.2 | XP_015644259.2 | 10 - 11 | 1180 | 118 | |
Set Membership: |
|