Assay Details
| Gene Symbol: LOC4340890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788773.2 | XP_015644259.2 | 12 - 13 | 1381 | 70 | |
| GenBank mRNA | AK241587.1 | - | 11 - 12 | 1481 | 70 | |
More Information
Set Membership: |
|

LOC4340890
Genomic Map