Assay Details
| Gene Symbol: LOC4340890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788773.2 | XP_015644259.2 | 7 - 8 | 808 | 129 | |
| GenBank mRNA | AK241587.1 | - | 7 - 8 | 818 | 129 | |
More Information
Set Membership: |
|

LOC4340890
Genomic Map