Assay Details
| Gene Symbol: LOC4337181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780399.2 | XP_015635885.1 | 9 - 10 | 1972 | 78 | |
| XM_015780400.2 | XP_015635886.1 | 8 - 9 | 1850 | 78 | ||
| XM_015780402.2 | XP_015635888.1 | 9 - 10 | 1949 | 78 | ||
| GenBank mRNA | AK105170.1 | - | 1 - 2 | 76 | 78 | |
More Information
Set Membership: |
|

LOC4337181
Genomic Map