Assay Details
| Gene Symbol: LOC4337396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778453.2 | XP_015633939.1 | 7 - 8 | 1172 | 74 | |
| GenBank mRNA | AK287813.1 | - | 7 - 8 | 1180 | 74 | |
| DQ269457.1 | - | 7 - 8 | 1098 | 74 | ||
More Information
Set Membership: |
|

LOC4337396
Genomic Map