Assay Details
| Gene Symbol: LOC4337788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782057.2 | XP_015637543.1 | 10 - 11 | 2303 | 78 | |
| GenBank mRNA | AK099724.1 | - | 10 - 11 | 2308 | 78 | |
More Information
Set Membership: |
|

LOC4337788
Genomic Map