Assay Details
| Gene Symbol: LOC4337788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782057.2 | XP_015637543.1 | 5 - 6 | 1821 | 123 | |
| GenBank mRNA | AK099724.1 | - | 5 - 6 | 1826 | 123 | |
More Information
Set Membership: |
|

LOC4337788
Genomic Map