Assay Details
| Gene Symbol: LOC4337945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782008.2 | XP_015637494.1 | 2 - 3 | 1077 | 88 | |
| XM_015782009.2 | XP_015637495.1 | 2 - 3 | 983 | 88 | ||
| GenBank mRNA | AK103471.1 | - | 2 - 3 | 981 | 88 | |
More Information
Set Membership: |
|

LOC4337945
Genomic Map